Order Entry
Belgium
ContactUsLinkComponent
 
Chlorobenzene ≥99%, GPR RECTAPUR®
 
undefined
Chlorobenzene ≥99%, GPR RECTAPUR®
 
 
 

 

  • Assay:
    Min. 99 %
  • Boiling point:
    131 - 133 °C
  • Density (20/4):
    1.100 - 1.110
  • Evaporation residue:
    Max. 50 ppm

 

Frequently Bought Together