Order Entry
Belgium
ContactUsLinkComponent
1,2,4-Trichlorobenzene ≥99% for HPLC, for GPC (gel permeation chromatography), J.T.Baker®
Irritant
Environment
1,2,4-Trichlorobenzene ≥99% for HPLC, for GPC (gel permeation chromatography), J.T.Baker®
Catalog # 9444-05
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Environment
1,2,4-Trichlorobenzene ≥99% for HPLC, for GPC (gel permeation chromatography), J.T.Baker®
Catalog # 9444-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    3,8 L
  • Seq:
    5138

Specifications

Specification Test Results

  • Assay (C₆H₃Cl₃) (by GC):
    ≥99 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:
    ≤0.15
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 375 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm:
    ≤0.010
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤310
  • Residue after Evaporation:
    ≤5 ppm
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.01 %

Specification Test Results

About this item

 :
 :181.45 g/mol
Merck Index:13,09704
Boiling Pt:214.4 °C (1013 hPa)
Melting Pt:17 °C
Density:1.45 g/cm³ (20 °C)
Flash Pt:99 °C
Storage Temperature:Ambient
EINECS:204-428-0
UN:2321
ADR:6.1,III
 :GHS07
 :GHS09
CAS Number:120-82-1
Formula:C₆H₃Cl₃
MDL Number:MFCD00000547