Orders LinkContactUsLinkComponent
Conductivity cells
Supplier:   SI Analytics
Conductivity cells
Supplier:  SI Analytics
Certificates
Conductivity cells
Conductivity cells
5 Options Available Below
Product Details & Documents
Robust conductivity cells for on-line measuring. Cells are either available with POM shaft or with glass shaft for high chemical resistance.
  • Screw plug head with thread Pg 13.5
  • LF 1101 versions and LF 1051 with platinum plates
  • LF 5101 versions and LF 71 T with platinum rings
  • HD electrode: Certified pressure and temperature consistency according to DIN EN 10204
Pressure range: 10 bar
 : All electrodes (except LF 1101 T HD) supplied with washer 9980/52 and O-ring type 9932/07. LF 1101 T HD is supplied with HD sealing type 9980/51. Connecting cables need to be ordered separately.
Product Family Options
Product Information
  • Type
    Conductivity
    Cell constant
    Temp.
    Material
    Ø×L
    Connection
    Availability
    Price
  • 662-1754
    2-pole conductivity cell
    2-pole conductivity cell
    LF 1101
    0 - 200 mS/cm
    1,0 cm-1
    −30...+100 °C
    Glass
    12×120 mm
    Screw plug head
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 586,00 €
    586,00 €
    1 * 1 ST
       586,00 €
  • 662-1755
    2-pole conductivity cell, with PT1000 temperature sensor
    2-pole conductivity cell, with PT1000 temperature sensor
    LF 1101 T
    0 - 200 mS/cm
    1,0 cm-1
    −30...+100 °C
    Glass
    12×120 mm
    SMEK screw plug head
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 863,00 €
    863,00 €
    1 * 1 ST
       863,00 €
  • 662-1756
    2-pole conductivity cell
    2-pole conductivity cell
    LF 5101
    0 - 200 mS/cm
    1,0 cm-1
    0...+80 °C
    POM
    12×120 mm
    Screw plug head
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 469,00 €
    469,00 €
    1 * 1 ST
       469,00 €
  • 662-1757
    2-pole conductivity cell, with PT1000 temperature sensor
    2-pole conductivity cell, with PT1000 temperature sensor
    LF 5101 T
    0 - 200 mS/cm
    1,0 cm-1
    0...+80 °C
    POM
    12×120 mm
    Screw plug head
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 606,00 €
    606,00 €
    1 * 1 ST
       606,00 €
  • SCOI285119720
    4-pole conductivity cell, with PT1000 temperature sensor
    4-pole conductivity cell, with PT1000 temperature sensor
    LF 71T1X
    0 - 200 mS/cm
    1,0 cm-1
    −30...+100 °C
    Glass
    12×120 mm
    fixed cable, open end
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €