Orders LinkContactUsLinkComponent
Automatic burettes, Pellet pattern, lateral stopcock
Automatic burettes, Pellet pattern, lateral stopcock
Supplier:   HIRSCHMANN
Automatic burettes, Pellet pattern, lateral stopcock
Supplier:  HIRSCHMANN
Certificates
Automatic burettes, Pellet pattern, lateral stopcock
Automatic burettes, Pellet pattern, lateral stopcock
6 Options Available Below
Product Details & Documents
Automatic burettes, Pellet pattern, class B, DURAN® with blue graduation.
  • Schellbach stripes for enhanced legibility
  • Lateral glass stopcock or PTFE valve stopcock
Certifications: Caliberated for DIN EN ISO 385.
Documents
Product Family Options
Lateral glass stopcock without reservoir bottle
Product Information
  • Capacity
    Division
    Scale length
    Tolerance
    Availability
    Price
  • 10 ml
    0,02 ml
    500 mm
    0,03 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 239,00 €
    239,00 €
    1 * 1 ST
       239,00 €
  • 25 ml
    0,05 ml
    500 mm
    0,045 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 239,00 €
    239,00 €
    1 * 1 ST
       239,00 €
  • 50 ml
    0,1 ml
    500 mm
    0,075 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 239,00 €
    239,00 €
    1 * 1 ST
       239,00 €
Lateral stopcock with PTFE spindle without reservoir bottle
Product Information
  • Capacity
    Division
    Scale length
    Tolerance
    Availability
    Price
  • 10 ml
    0,02 ml
    500 mm
    0,03 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 236,00 €
    236,00 €
    1 * 1 ST
       236,00 €
  • 25 ml
    0,05 ml
    500 mm
    0,045 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 236,00 €
    236,00 €
    1 * 1 ST
       236,00 €
  • 50 ml
    0,1 ml
    500 mm
    0,075 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 236,00 €
    236,00 €
    1 * 1 ST
       236,00 €
Reservoir bottle
Product Information
  • Catalog #
    Description
    Capacity
    Availability
    Price
  • Reservoir bottle, NS 29/32, DURAN® clear glass
    2000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,20 €
    31,20 €
    1 * 1 ST
       31,20 €