Orders LinkContactUsLinkComponent
GC septa, three layer
GC septa, three layer
Supplier:   HAMILTON BONADUZ
GC septa, three layer
Supplier:  HAMILTON BONADUZ
Certificates
GC septa, three layer
GC septa, three layer
15 Options Available Below
Product Details & Documents
These preconditioned silicone rubber septa are 3,81 mm thick.
  • Average life up to 100 injections with a 26s gauge needle
  • Maximum temperature up to 400 °C
Product Family Options
Product Information
  • Ø
    Thickness
    Availability
    Price
  • HAMI76022
    Three layer GC septa
    Three layer GC septa
    5 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 29,30 €
    29,30 €
    1 * 12 ST
       29,30 €
  • HAMI76001
    Three layer GC septa
    Three layer GC septa
    6,35 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 22,80 €
    22,80 €
    1 * 12 ST
       22,80 €
  • HAMI76010
    Three layer GC septa
    Three layer GC septa
    7 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 21,70 €
    21,70 €
    1 * 12 ST
       21,70 €
  • HAMI76002
    Three layer GC septa
    Three layer GC septa
    8 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 26,00 €
    26,00 €
    1 * 12 ST
       26,00 €
  • HAMI76003
    Three layer GC septa
    Three layer GC septa
    9 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 27,10 €
    27,10 €
    1 * 12 ST
       27,10 €
  • HAMI76004
    Three layer GC septa
    Three layer GC septa
    9,5 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 24,90 €
    24,90 €
    1 * 12 ST
       24,90 €
  • HAMI76005
    Three layer GC septa
    Three layer GC septa
    10 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 30,30 €
    30,30 €
    1 * 12 ST
       30,30 €
  • HAMI76007
    Three layer GC septa
    Three layer GC septa
    11 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 33,60 €
    33,60 €
    1 * 12 ST
       33,60 €
  • HAMI76013
    Three layer GC septa
    Three layer GC septa
    12 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 36,40 €
    36,40 €
    1 * 12 ST
       36,40 €
  • HAMI76006
    Three layer GC septa
    Three layer GC septa
    12,7 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 42,20 €
    42,20 €
    1 * 12 ST
       42,20 €
  • 613-1520
    Three layer GC septa
    Three layer GC septa
    13 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 40,10 €
    40,10 €
    1 * 12 ST
       40,10 €
  • HAMI76015
    Three layer GC septa
    Three layer GC septa
    14 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 52,00 €
    52,00 €
    1 * 12 ST
       52,00 €
  • HAMI76016
    Three layer GC septa
    Three layer GC septa
    15 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 45,50 €
    45,50 €
    1 * 12 ST
       45,50 €
  • HAMI76008
    Three layer GC septa
    Three layer GC septa
    16 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 29,30 €
    29,30 €
    1 * 12 ST
       29,30 €
  • HAMI76020
    Three layer GC septa
    Three layer GC septa
    19 mm
    3,81 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 74,70 €
    74,70 €
    1 * 12 ST
       74,70 €