Dropping funnels, cylindrical, without pressure balance, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear, with glass key.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape. With standard ground joint cone and socket. Clearly visible divisions for approximate volume reading.
DIN ISO 4800.
* non-DIN ISO size
Frequently Bought Together