Orders LinkContactUsLinkComponent
Conductivity cells, ScienceLine
Conductivity cells, ScienceLine
Supplier:   SI Analytics
Conductivity cells, ScienceLine
Supplier:  SI Analytics
Certificates
Conductivity cells, ScienceLine
Conductivity cells, ScienceLine
2 Options Available Below
Product Details & Documents
These high-end conductivity cells are ideal for the monitoring and control of plants as well as the examination of ultrapure water in quality control labs.
  • Models with SMEK plug head can be connected to diverse instrument types
  • Glass platinum electrodes LF713T (4-pole) and LF913T (5-pole) allow the determination of conductivity in organic media
Product Family Options
Product Information
  • Type
    Conductivity
    Cell constant
    Temp.
    Material
    Ø×L
    Connection
    Availability
    Price
  • 663-4006
    4-pole conductivity cell, sensor 4 platinum rings, temp. sensor NTC 30 kOhm
    4-pole conductivity cell, sensor 4 platinum rings, temp. sensor NTC 30 kOhm
    LF 613 T
    1 µS/cm - 2000 mS/cm
    1,0 cm-1
    −5...+80 °C
    Plastic
    12×120 mm
    Fixed 1 m cable with 8 pole plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 724,00 €
    724,00 €
    1 * 1 ST
       724,00 €
  • 663-0282
    4-pole conductivity cell, sensor 4 platinum rings, temp. sensor NTC 30 kOhm
    4-pole conductivity cell, sensor 4 platinum rings, temp. sensor NTC 30 kOhm
    LF 713 T
    1 µS/cm - 2000 mS/cm
    1,0 cm-1
    −30...+135 °C
    Glass
    12×120 mm
    Fixed 1 m cable with 8 pole plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 869,00 €
    869,00 €
    1 * 1 ST
       869,00 €