Orders LinkContactUsLinkComponent
SupaMega tissue processing cassette
SupaMega tissue processing cassette
White
Catalog # 720-1897
Supplier:   CELLPATH
SupaMega tissue processing cassette
Catalog # 720-1897
Supplier:  CELLPATH
SupaMega tissue processing cassette
SupaMega tissue processing cassette
White
Price
167,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
CellPath’s SupaMega Cassette is ideal for embedding and processing large tissue samples. The SupaMega Cassette is widely used in research and toxicology labratories. Dimension: 65×50×15 mm
  • Ideal for prostate, brain, breast tissue and eyes
  • Extremely useful for research and toxicology laboratories
Specifications
  • Colour:
    White