Orders LinkContactUsLinkComponent
Reference electrodes
Reference electrodes
Supplier:   SI Analytics
Reference electrodes
Supplier:  SI Analytics
Certificates
Reference electrodes
Reference electrodes
6 Options Available Below
Product Details & Documents
These reference electrodes are available with low maintenance plug heads or with fixed cables and free end.
Product Family Options
With plug head
Product Information
  • Type
    Temp.
    Material
    Electrolyte
    Diaphr.
    Ø×L
    Connection
    Availability
    Price
  • SCOI285111767
    Reference electrode, low maintenance, Ag/AgCl reference system
    Reference electrode, low maintenance, Ag/AgCl reference system
    B 1881 HD
    −5...+100 °C
    PVDF
    Paste
    PVDF
    12×120 mm
    Screw plug head
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 300,00 €
    300,00 €
    1 * 1 ST
       300,00 €
  • SCOI285111915
    Reference electrode, low maintenance, Silamid® reference system
    Reference electrode, low maintenance, Silamid® reference system
    B 1981 HD
    −5...+100 °C
    Glass
    Referid®
    KPG annular gap
    12×120 mm
    Screw plug head
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 320,00 €
    320,00 €
    1 * 1 ST
       320,00 €
  • SCOI285111701
    Reference electrode, low maintenance, Ag/AgCl reference system
    Reference electrode, low maintenance, Ag/AgCl reference system
    B 1181 HD
    −5...+50 °C
    Glass
    Paste
    Ceramic
    12×120 mm
    Screw plug head
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 237,00 €
    237,00 €
    1 * 1 ST
       237,00 €
  • SCOI285111504
    Reference electrode, Ag/AgCl reference system
    Reference electrode, Ag/AgCl reference system
    B 1491 HD
    +5...+135 °C
    Glass
    KCI 3 mol/l
    Ceramic
    12×170 mm
    Screw plug head
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 258,00 €
    258,00 €
    1 * 1 ST
       258,00 €
With fixed cable
Product Information
  • Type
    Temp.
    Material
    Electrolyte
    Diaphr.
    Ø×L
    Connection
    Availability
    Price
  • SCOI285116851
    Reference electrodes with free end, Ag/AgCl reference system and refilling tube
    Reference electrodes with free end, Ag/AgCl reference system and refilling tube
    9828/95
    +5...+135 °C
    Glass
    KCI 3 mol/l
    Ceramic
    12×120 mm
    5 m fixed cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 317,00 €
    317,00 €
    1 * 1 ST
       317,00 €
  • SCOI285117856
    Reference electrodes with free end, flanged cap, low maintatence, Ag/AgCl reference system
    Reference electrodes with free end, flanged cap, low maintatence, Ag/AgCl reference system
    9801/85
    −5...+50 °C
    Glass
    Paste
    Ceramic
    12×120 mm
    5 m fixed cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 236,00 €
    236,00 €
    1 * 1 ST
       236,00 €