Orders LinkContactUsLinkComponent
DURAN® Evaporating Flasks
DURAN® Evaporating Flasks
Supplier:   DWK Life Sciences
DURAN® Evaporating Flasks
Supplier:  DWK Life Sciences
Certificates
DURAN® Evaporating Flasks
DURAN® Evaporating Flasks
4 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard rotary evaporators; with white labelling area.
Product Family Options
Product Information
  • Capacity
    Socket size
    Total height
    Availability
    Price
  • 100 ml
    29/32 NS
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,00 €
    42,00 €
    1 * 1 ST
       42,00 €
  • 250 ml
    29/32 NS
    140 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 47,90 €
    47,90 €
    1 * 1 ST
       47,90 €
  • 500 ml
    29/32 NS
    170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,90 €
    52,90 €
    1 * 1 ST
       52,90 €
  • 1000 ml
    29/32 NS
    210 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,50 €
    70,50 €
    1 * 1 ST
       70,50 €