Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium spoons

Sodium spoons

Supplier:   USBECK Laborgeräte
Sodium spoons
Certificates
Sodium spoons

2 Options Available Below

Product Details & Documents

Copper basket.

  • With ergonomic handle
  • Single or double
  • Basket Ø 25 mm
Product Family Options

Product Information

  • Length
    Availability
    Price
  • 231-1271
    Sodium spoon, single, 16 g
    Sodium spoon, single, 16 g
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,70 €
    10,70 €
    1 * 1 ST
       10,70 €

  • USBE3412
    Sodium spoon, double, 26 g
    Sodium spoon, double, 26 g
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,20 €
    19,20 €
    1 * 1 ST
       19,20 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.