Orders LinkContactUsLinkComponent
Micro filter funnels, DURAN®
Micro filter funnels, DURAN®
Supplier:   DWK Life Sciences
Micro filter funnels, DURAN®
Supplier:  DWK Life Sciences
Certificates
Micro filter funnels, DURAN®
Micro filter funnels, DURAN®
4 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • For filtration of very small volumes
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Very good temperature resistance
With integrated sintered glass filter plate.
Product Family Options
Product Information
  • Plate Ø
    Porosity
    Capacity
    Stem O-Ø
    Availability
    Price
  • 10 mm
    1
    2 ml
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,00 €
    46,00 €
    1 * 1 ST
       46,00 €
  • 10 mm
    2
    2 ml
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,00 €
    46,00 €
    1 * 1 ST
       46,00 €
  • 10 mm
    3
    2 ml
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,00 €
    46,00 €
    1 * 1 ST
       46,00 €
  • 10 mm
    4
    2 ml
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,10 €
    49,10 €
    1 * 1 ST
       49,10 €