ORP standards for Redox/ORP electrodes, Orion™
Supplier: Thermo Orion
Certificates
About this item
Measure ORP simply and accurately with Thermo Scientific™ Orion™ ORP standards for Redox/ORP electrodes
- Does not contain hazardous chemicals
- Included in the new ASTM ORP method
- Used as relative mV standard to calibrate the offset of sample
Orion™ OPR standards are designed for use with ORP/ATC Triode™ electrodes and Sure-Flow™ ORP electrodes.
Stable for up to a year if unopened and can be used directly from the bottle with no mixing or diluting.
Frequently Bought Together