4-Aminobenzoic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:150-13-0
:
Formula:C₇H₇NO₂
MDL Number:MFCD00007894
:137.14 g/mol
Merck Index:13,00422
Boiling Pt:340 °C (1013 hPa)
Melting Pt:188 °C
Density:1.45 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:205-753-0
Specification Test Results
- Solubility:4.7 g/l (20 °C)
- Melting Point:186 - 189 °C
- Molar Mass:137.14 g/mol
- Bulk Density:200 kg/m3
- Density:1.38 g/cm3
- pH:3.5 (5 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Frequently Bought Together