Pocket refractometer, DEF-Chek
Digital tester for urea concentration (%) in Diesel Exhaust Fluid (DEF), DEF-Chek
Catalog # 635-0507
Supplier: REICHERT
Jump to:
Product Details & Documents
Selective catalytic reduction systems for diesel trucks and buses requires Diesel Exhaust Fluid (DEF) at an exact concentration of 32,5 % urea.
The DEF-Chek offers quick accurate results.
Specifications
- Model:DEF-Chek
- Description:Digital tester for urea concentration (%) in Diesel Exhaust Fluid (DEF)
- Seq:003
Accessories
Product Information
- Catalog #AvailabilityPrice
- Nylon holster case for Chek series