pH combination electrodes with temperature sensor, ScienceLine
Supplier: SI Analytics
pH combination electrodes with temperature sensor, ScienceLine
Supplier: SI Analytics
Certificates
Jump to:
About this item
Electrodes with plug head (without cable) or with BNC/DIN plug with fixed cable.
- Reference system: Silamid®
- Membrane shape: sphere
- Electrodes with suffix 'ID' feature an ID function (electrode data is stored in the connector for the automatic sensor recognition)
7 Options Available Below
Product Details & Documents
Electrodes with plug head (without cable) or with BNC/DIN plug with fixed cable.
- Reference system: Silamid®
- Membrane shape: sphere
- Electrodes with suffix 'ID' feature an ID function (electrode data is stored in the connector for the automatic sensor recognition)
Documents
Product Family Options
Product Information
- TypepHTemp.MaterialElectrolyteDiaphr.Ø×LConnectionAvailabilityPrice
- 661-0243Combination electrode, sphere membrane, A-glass, with PT1000 temperature sensor, Silamid®-reference system, with sensor recognitionCombination electrode, sphere membrane, A-glass, with PT1000 temperature sensor, Silamid®-reference system, with sensor recognitionA 162 2M-DIN-ID0 - 14−5...+100 °CGlassKCl 3 mol/lPlatinum12×120 mmDIN + 4 mm banana plug, 2 m fixed cableLog in for availability1 * 1 ST474,00 €
- 662-1972Combination electrode, sphere membrane, A-glass, with PT1000 temperature sensor, Silamid®-reference system, with sensor recognitionCombination electrode, sphere membrane, A-glass, with PT1000 temperature sensor, Silamid®-reference system, with sensor recognitionA 7780 1M-DIN-ID0 - 14−5...+80 °CGlassGel3×Ceramic12×120 mmDIN + 4 mm banana plug, 1 m fixed cableLog in for availability1 * 1 ST462,00 €
- 662-1377Combination electrode, sphere membrane, A-glass, with NTC 30 kOhm temperature sensor, Silamid®-reference systemCombination electrode, sphere membrane, A-glass, with NTC 30 kOhm temperature sensor, Silamid®-reference systemA 7780 NTC30 DIN N0 - 14−5...+80 °CGlassGel3×Ceramic12×120 mmDIN + 4 mm banana plug, 1 m fixed cableLog in for availability1 * 1 ST382,00 €
- 662-6111Combination electrode, sphere membrane, A-glass, with PT1000 temperature sensor, Silamid®-reference systemCombination electrode, sphere membrane, A-glass, with PT1000 temperature sensor, Silamid®-reference systemN 1041 A0 - 14−5...+100 °CGlassKCl 3 mol/lCeramic12×170 mmDIN + 4 mm banana plug, 1 m fixed cableLog in for availability1 * 1 ST533,00 €
- 662-6106Combination electrode, sphere membrane, A-glass, with PT1000 temperature sensor, Silamid®-reference systemCombination electrode, sphere membrane, A-glass, with PT1000 temperature sensor, Silamid®-reference systemN 1042 A0 - 14−5...+100 °CGlassKCl 3 mol/lCeramic12×120 mmDIN + 4 mm banana plug, 1 m fixed cableLog in for availability1 * 1 ST523,00 €
- 662-1941Combination electrode, sphere membrane, with PT1000 temperature sensor, Silamid®-reference systemCombination electrode, sphere membrane, with PT1000 temperature sensor, Silamid®-reference systemN 1043 A0 - 14−5...+100 °CGlassKCl 3 mol/lCeramic12×320 mmDIN + 4 mm banana plug, 1 m fixed cableLog in for availability1 * 1 ST589,00 €
- 662-1949Combination electrode, sphere membrane, A-glass, with PT1000 temperature sensor, Silamid®-reference systemCombination electrode, sphere membrane, A-glass, with PT1000 temperature sensor, Silamid®-reference systemN 1051 A0 - 14−5...+100 °CGlassKCl 3 mol/lPlatinum12×170 mmDIN + 4 mm banana plug, 1 m fixed cableLog in for availability1 * 1 ST539,00 €