Benzoic acid, Certipur® Reag. USP, Reag. Ph. Eur. volumetric standard, secondary reference material for alkalimetry, traceable to NIST Standard Reference Material (SRM), Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:65-85-0
:
Formula:C₇H₆O₂
MDL Number:MFCD00002398
:122.12 g/mol
Merck Index:13,01092
Boiling Pt:249 °C (1013 hPa)
Melting Pt:121…123 °C
Density:1.316 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:121 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-618-2
REACH:01-2119455536-33
:GHS05
:GHS08
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:2.9 g/l (25 °C)
- Melting Point:121 - 123 °C
- Molar Mass:122.12 g/mol
- Bulk Density:500 kg/m3
- Boiling Point:249 °C (1013 hPa)
- Vapor Pressure:0.001 hPa (20 °C)
- Flash Point:121 °C
- Density:1.321 g/cm3 (20 °C)
- pH:3.1 (1 g/l, H2O, 20 °C)
- Ignition Point:570 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberCertipur Merck Brighter titration Titripur® Merck Segment Brochure Merck Supelco® Titration goes Digital
Product Literature
msds_102401
SDS
ProductPage_102401
Product specification
Frequently Bought Together