Rotating Sample Reducer, RPT
Supplier: VWR Collection
Jump to:
About this item
The HAVER rotating sample reducer produces small but representative sample quantities of solids and suspensions. It combines three dividing methods in one unit and achieves an accuracy of 99,9%.
- Representative sample division at a rate of 1:8, 1:10 or 1:30
- Integrated adjustable feeder with continuously regulated vibration intensity
- Easier material flow caused by high rotating speed of the cone (approx. 100 U/min.)
- Made from food grade materials - easy to clean
- Fitted with a low maintenance drive motor with safety clutch
3 Options Available Below
Product Details & Documents
The HAVER rotating sample reducer produces small but representative sample quantities of solids and suspensions. It combines three dividing methods in one unit and achieves an accuracy of 99,9%.
- Representative sample division at a rate of 1:8, 1:10 or 1:30
- Integrated adjustable feeder with continuously regulated vibration intensity
- Easier material flow caused by high rotating speed of the cone (approx. 100 U/min.)
- Made from food grade materials - easy to clean
- Fitted with a low maintenance drive motor with safety clutch
Product Family Options
Product Information
- Max. feed quantityMax. feed sizeAvailabilityPrice
- HAVE205322419Rotating sample reducer with 30 funnels, outlets and 30 flasks (15 ml)Rotating sample reducer with 30 funnels, outlets and 30 flasks (15 ml)300 ml2,5 mmLog in for availability1 * 1 ST12 100,00 €Specifications:
More
Specifications
Cat. No.HAVE205322419 - HAVE205322402Rotating sample reducer with 10 funnels, outlets and 10 flasks (250 ml)Rotating sample reducer with 10 funnels, outlets and 10 flasks (250 ml)2500 ml10 mmLog in for availability1 * 1 ST10 240,00 €Specifications:
More
Specifications
Cat. No.HAVE205322402 - HAVE205322396Rotating sample reducer with 8 funnels, outlets and 8 flasks (500 ml)Rotating sample reducer with 8 funnels, outlets and 8 flasks (500 ml)4000 ml10 mmLog in for availability1 * 1 ST10 200,00 €Specifications:
More
Specifications
Cat. No.HAVE205322396