p-Anisaldehyde ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:123-11-5
:
Formula:C₈H₈O₂
MDL Number:MFCD00003385
Merck Index:13,00666
Boiling Pt:247…249 °C (1013 hPa)
Melting Pt:0…2 °C
Density:1.12 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:110 °C
:136.15 g/mol
Storage Temperature:Ambient
EINECS:204-602-6
Specification Test Results
- Solubility:2 g/l (20 °C)
- Molar Mass:136.15 g/mol
- Boiling Point:247 - 249 °C (1013 hPa)
- Vapor Pressure:- 1 hPa (20 °C)
- Flash Point:116 °C
- Refractive Index:1.57 (20 °C)
- Explosion Limit:1.4 - 5.3 %(V)
- Density:1.12 g/cm3 (25 °C)
- pH:7 (2 g/l, H2O, 20 °C)
- Ignition Point:220 °C
Specification Test Results
Related Information
Frequently Bought Together