ContactUsLinkComponent
Mechanical overhead stirrers, RKG-00-Bo, TURBOTRON®, IKA®
Mechanical overhead stirrers, RKG-00-Bo, TURBOTRON®, IKA®
Supplier:  IKA
Certificates
Mechanical overhead stirrers, RKG-00-Bo, TURBOTRON®, IKA®
Supplier:  IKA
Specifications
  • Catalog No:
  • IKAAU098950
  • IKAAU084463
  • IKAAU087228
  • Model:
  • RKG-00-Bo ATEX
  • RKG-00-Bo ATEX
  • RKG-00-Bo ATEX
  • Max. viscosity:
  • 50000 mPas
  • 50000 mPas
  • 100000 mPas
  • Speed range I:
  • 20 - 1200 min⁻¹
  • 20 - 1200 min⁻¹
  • 20 - 1200 min⁻¹
  • Max. torque:
  • ~300 Ncm at 20 - 600 min⁻¹
    ~150 Ncm at 1200 min⁻¹
  • ~300 Ncm at 20 - 600 min⁻¹
    ~150 Ncm at 1200 min⁻¹
  • ~600 Ncm at 20 - 600 min⁻¹
    ~330 Ncm at 1200 min⁻¹
  • W×D×H:
  • 369×288×537 mm
  • 369×288×537 mm
  • 196×333×550 mm
  • Weight:
  • 13,8 kg
  • 13,8 kg
  • 20,0 kg