Orders LinkContactUsLinkComponent
Staining jars, DURAN®
Staining jars, DURAN®
Staining dishes acc. to Schiefferdecker, for up to 10 slides, 70×90×40 mm
  SCOT233160003
Staining jars, DURAN®
  SCOT233160003
Staining jars, DURAN®
Staining jars, DURAN®
Staining dishes acc. to Schiefferdecker, for up to 10 slides, 70×90×40 mm
241,00 €
1 * 10 ST
Soda-lime glass.
  • For 10 slides
These staining jars are suitable for standard microscope slides (76×26 mm).
 : Note: Do not clean staining dishes and jars at temperatures above 60°C (glass corrosion is possible).
  • Description:
    Staining dishes acc. to Schiefferdecker, for up to 10 slides
  • W×D×H:
    70×90×40 mm