
Ethanol denatured ≥96% (v/v), TechniSolv® (denatured with 3% isopropanol)
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64-17-5
:
Formula:H₃CCH₂OH
MDL Number:MFCD00003568
:46.07 g/mol
Density:0.789 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Assay (Ethanol) (V/V):Min. 96 %
- Propan-2-ol (V/V):Min. 3 %
- Water:Max. 0.5 %
Specification Test Results
Related Information
Certificates
Search by Lot Number25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
87137_ETHANOL 99 % DENATURED WITH 3% IPA (VV)_BSE_REV0
BSE/TSE Certificates
spec_VWRC87137.360 spec_VWRC87137.360 spec_VWRC87137.460 spec_VWRC87137.460
Product specification
Frequently Bought Together