ContactUsLinkComponent
Penicillin flask, DURAN®
Penicillin flask, DURAN®
Catalog # 391-0370
Supplier:  DWK Life Sciences
undefined
Penicillin flask, DURAN®
Catalog # 391-0370
Supplier:  DWK Life Sciences
Specifications
  • Capacity:
    4000 ml
  • Neck Ø ext.:
    50 mm
  • Pk:
    1
Specifications
About this item
This penicillin flask is made from clear DURAN® borosilicate glass 3.3.
  • Neck is bent upwards which prevents the liquid culture medium from leaking
  • Due to the flat, large floor surface, the layer cover of microorganisms can grow evenly high
  • The large liquid surface ensures better aeration and thus faster growth of the microorganisms
Typically suitable for preparation of cultures in nutrient media.