Vanillin 99%, pure
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 4-Hydroxy-3-methoxybenzaldehyde
- CAS Number:121-33-5
- Formula:C₈H₈O₃
- MDL Number:MFCD00006942
- Molecular Weight:152.15 g/mol
- Merck Index:13,09998
- Boiling Pt:170 °C (15 mmHg)
- Melting Pt:81…84 °C
- Density:1,056 g/cm³ (20 °C)
- Flash Pt:147 °C
- Storage Temperature:Ambient
- EINECS:204-465-2
5 Options Available Below
Product Details & Documents
Synonyms: 4-Hydroxy-3-methoxybenzaldehyde
- CAS Number:121-33-5
- Formula:C₈H₈O₃
- MDL Number:MFCD00006942
- Molecular Weight:152.15 g/mol
- Merck Index:13,09998
- Boiling Pt:170 °C (15 mmHg)
- Melting Pt:81…84 °C
- Density:1,056 g/cm³ (20 °C)
- Flash Pt:147 °C
- Storage Temperature:Ambient
- EINECS:204-465-2
Specifications for Product Family
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:81 to 84 °C
- Titration with NaOH:≥98.5 %
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



