Orders LinkContactUsLinkComponent
NMR sample tubes, 5 mm, with accuracy classes, DURAN®
NMR sample tubes, 5 mm, with accuracy classes, DURAN®
Supplier:   DWK Life Sciences
NMR sample tubes, 5 mm, with accuracy classes, DURAN®
Supplier:  DWK Life Sciences
Certificates
NMR sample tubes, 5 mm, with accuracy classes, DURAN®
NMR sample tubes, 5 mm, with accuracy classes, DURAN®
3 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Storage and transport of measurement solutions
  • Designed for use in low to most mid field NMR spectrometers
  • Close tolerances and accuracy, especially to their straightness, wall thickness and wall thickness distribution
DURAN® NMR tubes are available, according to requirement, in three accuracy classes. The correct tube can be selected depending on resonant frequency. These tubes are noteworthy for their close tolerances and accuracy, especially to their straightness, wall thickness and wall thickness distribution. Consequently, quick and accurate test results are achievable.
With printed retrace code (batch identification).
Product Family Options
Product Information
  • Length
    Camber
    Frequency
    Availability
    Price
  • 178 mm
    70 µm
    300 MHz
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 191,00 €
    191,00 €
    1 * 50 ST
       191,00 €
  • 178 mm
    30 µm
    400 MHz
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 3 110,00 €
    3 110,00 €
    1 * 250 ST
       3 110,00 €
  • 178 mm
    130 µm
    500 MHz
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 116,00 €
    116,00 €
    1 * 5 ST
       116,00 €