Orders LinkContactUsLinkComponent
Extractor thimbles for solids
Extractor thimbles for solids
Supplier:   Lenz Laborglas
Extractor thimbles for solids
Supplier:  Lenz Laborglas
Certificates
Extractor thimbles for solids
Extractor thimbles for solids
3 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For glass inserts for solids.
Documents
Product Family Options
Product Information
  • Extraction cap.
    Socket size
    Availability
    Price
  • 70 ml
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €
  • 100 ml
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 119,00 €
    119,00 €
    1 * 1 ST
       119,00 €
  • 250 ml
    60/46 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 174,00 €
    174,00 €
    1 * 1 ST
       174,00 €