Orders LinkContactUsLinkComponent
Distilling receivers, Bernauer
Supplier:   Lenz Laborglas
Distilling receivers, Bernauer
Supplier:  Lenz Laborglas
Certificates
Distilling receivers, Bernauer
Distilling receivers, Bernauer
6 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling receivers acc. to Bernauer (modified), with three cones NS 14/23, vacuum connection with glass olive. With flasks 100 ml capacity or without flasks available.
Product Family Options
With flasks
Product Information
  • Socket size
    Availability
    Price
  • 14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 130,00 €
    130,00 €
    1 * 1 ST
       130,00 €
  • 19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 131,00 €
    131,00 €
    1 * 1 ST
       131,00 €
  • 29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 128,00 €
    128,00 €
    1 * 1 ST
       128,00 €
Without flasks
Product Information
  • Socket size
    Availability
    Price
  • 14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 97,10 €
    97,10 €
    1 * 1 ST
       97,10 €
  • 19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,90 €
    92,90 €
    1 * 1 ST
       92,90 €
  • 29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 99,20 €
    99,20 €
    1 * 1 ST
       99,20 €