Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Sodium heptadecafluorooctane-1-sulphonate 50 µg/ml in methanol
Organic reference standard, Sodium heptadecafluorooctane-1-sulphonate 50 µg/ml in methanol
Catalog # MSPPL-PFOS
CAS Number:  
Organic reference standard, Sodium heptadecafluorooctane-1-sulphonate 50 µg/ml in methanol
Catalog # MSPPL-PFOS
CAS Number:  

Specifications

  • Pk:
    1 Ampoul
  • CAS No.:
    4021-47-0
  • Description:
    Sodium heptadecafluorooctane-1-sulphonate 50 µg/ml in methanol
  • Formula:
    C₈F₁₇NaO₃S
  • Description:
    Sodium heptadecafluorooctane-1-sulphonate

Specifications

About this item