
3-Bromo-4-fluorobenzoic acid
Catalog # MOLE26314892-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.01 g/mol
EINECS:213-751-6
:GHS07
CAS Number:1007-16-5
Formula:C₇H₄BrFO₂
MDL Number:MFCD00042463