Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Bisacrylamide

Irritant
Marketsource
Supplier:   Serva
Bisacrylamide
Supplier:  Serva
Marketsource
Certificates
Bisacrylamide

2 Options Available Below

Product Details & Documents
Synonyms:  Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
  • CAS Number:110-26-9
  • Formula:(H₂C=CHCONH)₂CH₂
  • MDL Number:MFCD00008625
  • Molecular Weight:154.17 g/mol
  • Boiling Pt:125 °C (3.33 hPa)
  • Melting Pt:185 °C
  • Density:1.235 g/cm³ (20 °C)
  • Flash Pt:138 °C
  • Storage Temperature:Refrigerator
  • EINECS:203-750-9
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

    Specifications:
    Marketsource
    Marketsource
  • 50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g

    Specifications:
    Marketsource
    Marketsource