Order Entry
Belgium
ContactUsLinkComponent
Perchloric acid 0.1 mol/l (0.1 N) in aqueous solution volumetric solution
Oxidizing
Irritant
Perchloric acid 0.1 mol/l (0.1 N) in aqueous solution volumetric solution
Catalog # REGEP2010H
Supplier:  REAGECON
CAS Number:  
undefined
Oxidizing
Irritant
Perchloric acid 0.1 mol/l (0.1 N) in aqueous solution volumetric solution
Catalog # REGEP2010H
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :100.46 g/mol
Storage Temperature:Ambient
EINECS:231-512-4
UN:1802
ADR:8,II
 :GHS03
 :GHS07
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325