Benzophenone oxime
Glass bottle for solids, 25 g
Catalog # MOLE18663882-25G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:574-66-3
- Formula:C₁₃H₁₁NO
- MDL Number:MFCD00051461
- Molecular Weight:197.24 g/mol
- EINECS:209-373-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



