Order Entry
Belgium
ContactUsLinkComponent
Trilithium citrate tetrahydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Irritant
Trilithium citrate tetrahydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Catalog # 1.05683.9024
CAS Number:  
Irritant
Trilithium citrate tetrahydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Catalog # 1.05683.9024
CAS Number:  

Specifications

  • Pk:
    20 kg
  • Seq:
    4241

Specifications

About this item

 :
 :281.99 g/mol
EINECS:213-045-8
 :GHS07
Storage Temperature:Ambient
CAS Number:6080-58-6
Formula:C₆H₅Li₃O₇·4H₂O
MDL Number:MFCD00150030