Order Entry
Belgium
ContactUsLinkComponent
tri-Calcium dicitrate tetrahydrate, EMPROVE® ESSENTIAL USP, E333, FCC, DAC suitable for use as excipient, SAFC®
tri-Calcium dicitrate tetrahydrate, EMPROVE® ESSENTIAL USP, E333, FCC, DAC suitable for use as excipient, SAFC®
Catalog # 1.02092.9025
CAS Number:  
tri-Calcium dicitrate tetrahydrate, EMPROVE® ESSENTIAL USP, E333, FCC, DAC suitable for use as excipient, SAFC®
Catalog # 1.02092.9025
CAS Number:  

Specifications

  • Pack type:
    Cardboard carton
  • Pk:
    25 kg
  • Seq:
    4283
  • Environmentally Preferable:

Specifications

About this item

Synonyms: tri-Calcium dicitrate tetrahydrate, Calcium citrate tribasic tetrahydrate
 :
 :570.5 g/mol
EINECS:212-391-7
CAS Number:5785-44-4
Formula:C₁₂H₁₀Ca₃O₁₄·4H₂O
MDL Number:MFCD00150786