Magnesium citrate hydrate, anhydrous
Catalog # LOHM163437.9050
Supplier: LOHMANN
CAS Number:
Specifications
- Pk:50 kg
- Seq:4418
Specifications
About this item
:
:451.13 g/mol (anhy)
EINECS:222-093-9
CAS Number:3344-18-1
Formula:C₁₂H₁₀Mg₃O₁₄·xH₂O
MDL Number:MFCD09953657