Calcium citrate tetrahydrate
Catalog # LOHM162417.9025
Supplier: LOHMANN
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:570.5 g/mol
EINECS:212-391-7
CAS Number:5785-44-4
Formula:C₁₂H₁₀Ca₃O₁₄·4H₂O
MDL Number:MFCD00150786