
Sulphuric acid 5 mol/l (10 N) in aqueous solution
Catalog # HACH93153
Supplier: Hach
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589