Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid 0.8 mol/l (1.6 N) in aqueous solution
Irritant
Sulphuric acid 0.8 mol/l (1.6 N) in aqueous solution
Catalog # HACH1438901
Supplier:  Hach
CAS Number:  
Irritant
Sulphuric acid 0.8 mol/l (1.6 N) in aqueous solution
Catalog # HACH1438901
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS07
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589