
Sulphuric acid (1:3) (v/v) 38% (v/v), extra pure
:7664-93-9
:
:H₂SO₄
:MFCD00064589
:98.08 g/mol
:∼100 °C (1013 hPa)
:1.1…1.4 g/cm³ (20 °C)
:Ambient
:231-639-5
:2796
:8,II
:GHS05
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Bottles, Wide Neck, with Screw Cap
: VWR Collection
Sulphanilamide ≥99%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Bioindicator, Sterikon® plus
: Merck
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection