Order Entry
Belgium
ContactUsLinkComponent
Pesticide reference standard, 1,2,4,5-Tetrachlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, 1,2,4,5-Tetrachlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17354500CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 1,2,4,5-Tetrachlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17354500CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    95-94-3
  • Description:
    1,2,4,5-Tetrachlorobenzene 10 µg/ml in cyclohexane
  • Environmentally Preferable:
  • Formula:
    C₆H₂Cl₄
  • Description:
    1,2,4,5-Tetrachlorobenzene

Specifications

About this item