Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid ≥75% (v/v) in aqueous solution
 
Sulphuric acid ≥75% (v/v) in aqueous solution
  42555.K2
 :  
 
Sulphuric acid ≥75% (v/v) in aqueous solution
  42555.K2
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

 :
 :98.08 g/mol
 :Ambient
 :231-639-5
 :1830
 :8,II
 :GHS05
 :7664-93-9
 :H₂SO₄
 :MFCD00064589