Orders LinkContactUsLinkComponent
Separating funnels, spherical, DURAN®
Separating funnels, spherical, DURAN®
Supplier:   DWK Life Sciences
Separating funnels, spherical, DURAN®
Supplier:  DWK Life Sciences
Certificates
Separating funnels, spherical, DURAN®
Separating funnels, spherical, DURAN®
6 Options Available Below
Product Details & Documents
DURAN® borosilicate glass 3.3, clear, stopcock with glass key and plastic stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 193,00 €
    193,00 €
    1 * 1 ST
       193,00 €
  • 500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 232,00 €
    232,00 €
    1 * 1 ST
       232,00 €
  • 1000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 266,00 €
    266,00 €
    1 * 1 ST
       266,00 €
  • 2500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 373,00 €
    373,00 €
    1 * 1 ST
       373,00 €
  • 5000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 572,00 €
    572,00 €
    1 * 1 ST
       572,00 €
  • 10000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 764,00 €
    764,00 €
    1 * 1 ST
       764,00 €