Orders LinkContactUsLinkComponent
Measuring beakers with handle
Measuring beakers with handle
 :   KARTELL
Measuring beakers with handle
 :  KARTELL
Measuring beakers with handle
Measuring beakers with handle
PP, translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use and a wide variety of other applications. With embossed scale and spout.
Product Family Options
  • Capacity
    Height
    Ext.Ø
    Graduation Interval
  • 250 ml
    119 mm
    73 mm
    5 ml
    1 * 1 ST
    1 * 1 ST - 12,20 €
    12,20 €
    1 * 1 ST
       12,20 €
  • 500 ml
    140 mm
    93 mm
    10 ml
    1 * 1 ST
    1 * 1 ST - 13,80 €
    13,80 €
    1 * 1 ST
       13,80 €
  • 1000 ml
    169 mm
    117 mm
    10 ml
    1 * 1 ST
    1 * 1 ST - 18,90 €
    18,90 €
    1 * 1 ST
       18,90 €
  • 2000 ml
    213 mm
    149 mm
    20 ml
    1 * 1 ST
    1 * 1 ST - 39,10 €
    39,10 €
    1 * 1 ST
       39,10 €
  • 3000 ml
    240 mm
    172 mm
    50 ml
    1 * 1 ST
    1 * 1 ST - 53,70 €
    53,70 €
    1 * 1 ST
       53,70 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.