Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Dropper Caps
Closure for dropping bottles, 302 series
Closure for dropping bottles, 302 series
Closure for dropping bottles, 302 series
Catalog # 215-2471
Supplier:
Kautex
Expand image
Closure for dropping bottles, 302 series
Catalog # 215-2471
Supplier:
Kautex
Specifications
Thread:
18 mm
Description:
Combination closure with screw-on dropping tip, HDPE
Pk:
1
For bottles:
10/25/50 ml
Specifications
About this item
Product Details
Documentation
Product Details
Documentation