Orders LinkContactUsLinkComponent
Filtration funnels, conical
Filtration funnels, conical
Supplier:   Lenz Laborglas
Filtration funnels, conical
Supplier:  Lenz Laborglas
Certificates
Filtration funnels, conical
Filtration funnels, conical
2 Options Available Below
Product Details & Documents
Funnels, made of boroslicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filter plate.
Documents
Product Family Options
Product Information
  • Capacity
    Length
    Plate Ø
    Porosity
    Availability
    Price
  • LENZ07200325
    Conical filter funnel
    Conical filter funnel
    25 ml
    100 mm
    25 mm
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,30 €
    28,30 €
    1 * 1 ST
       28,30 €
  • LENZ07200425
    Conical filter funnel
    Conical filter funnel
    25 ml
    100 mm
    25 mm
    4
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,50 €
    31,50 €
    1 * 1 ST
       31,50 €