Orders LinkContactUsLinkComponent
Syringes, SampleLock™
Supplier:   HAMILTON BONADUZ
Syringes, SampleLock™
Supplier:  HAMILTON BONADUZ
Certificates
Syringes, SampleLock™
12 Options Available Below
Product Details & Documents
These syringes allow the collection, storage, transport and analysis of liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
  • Not autoclavable, gas sterilisable
  • An easy-to-use twist valve
  • A positive rear stop prevents loss of sample and plunger blowout
  • Fluid path includes SST, borosilicate glass and PTFE
The sample lock termination incorporates an on/off syringe valve with removable needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseous samples for GC analysis, and sample spiking.
Optional male luer and female luer lock adapters (sold separately) threads onto the SampleLock™ valve, making the syringes compatible with a multitude of connectors and fittings.
Product Family Options
1000 SampleLock™ syringes
Product Information
  • Gauge
    Volume
    Length
    Type of tip
    Type
    Availability
    Price
  • 22
    1000 µl
    51 mm
    2
    1001 RN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 317,00 €
    317,00 €
    1 * 1 ST
       317,00 €
  • 22
    2500 µl
    51 mm
    2
    1002 RN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 340,00 €
    340,00 €
    1 * 1 ST
       340,00 €
  • 22
    5000 µl
    51 mm
    2
    1005 RN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 406,00 €
    406,00 €
    1 * 1 ST
       406,00 €
  • 22
    10000 µl
    51 mm
    2
    1010 RN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 414,00 €
    414,00 €
    1 * 1 ST
       414,00 €
  • 22
    25000 µl
    51 mm
    2
    1025 RN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €
  • 22
    50000 µl
    51 mm
    2
    1050 RN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 884,00 €
    884,00 €
    1 * 1 ST
       884,00 €
  • 22
    100000 µl
    51 mm
    2
    1100 RN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 020,00 €
    1 020,00 €
    1 * 1 ST
       1 020,00 €
1700 SampleLock™ syringes
Product Information
  • Gauge
    Volume
    Length
    Type of tip
    Type
    Availability
    Price
  • 22s
    50 µl
    51 mm
    2
    1705 RN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 229,00 €
    229,00 €
    1 * 1 ST
       229,00 €
  • 22s
    100 µl
    51 mm
    2
    1710 RN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,00 €
    223,00 €
    1 * 1 ST
       223,00 €
  • 22s
    250 µl
    51 mm
    2
    1725 RN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 262,00 €
    262,00 €
    1 * 1 ST
       262,00 €
  • 22
    500 µl
    51 mm
    2
    1750 RN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 262,00 €
    262,00 €
    1 * 1 ST
       262,00 €
  • 22s
    250 µl
    51 mm
    2
    1725 RN calibrated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 508,00 €
    508,00 €
    1 * 1 ST
       508,00 €
Replacement needles
Product Information
  • Catalog #
    Type of tip
    Ø int.
    Gauge
    Length
    Ext.Ø
    Availability
    Price
  • 2
    0,41 mm
    22
    51 mm
    0,72 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 83,30 €
    83,30 €
    1 * 6 ST
       83,30 €
Product Information
  • Catalog #
    Type of tip
    Ø int.
    Gauge
    Length
    Ext.Ø
    Availability
    Price
  • 3
    0,41 mm
    22
    51 mm
    0,72 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 63,90 €
    63,90 €
    1 * 6 ST
       63,90 €
Product Information
  • Catalog #
    Type of tip
    Ø int.
    Gauge
    Length
    Ext.Ø
    Availability
    Price
  • 2
    0,15 mm
    22s
    51 mm
    0,72 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 83,30 €
    83,30 €
    1 * 6 ST
       83,30 €
Product Information
  • Catalog #
    Type of tip
    Ø int.
    Gauge
    Length
    Ext.Ø
    Availability
    Price
  • 3
    0,15 mm
    22s
    51 mm
    0,72 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 66,00 €
    66,00 €
    1 * 6 ST
       66,00 €
Product Information
  • Catalog #
    Type of tip
    Ø int.
    Gauge
    Length
    Ext.Ø
    Availability
    Price
  • 5
    0,15 mm
    22s
    51 mm
    0,72 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 246,00 €
    246,00 €
    1 * 6 ST
       246,00 €
Product Information
  • Catalog #
    Type of tip
    Ø int.
    Gauge
    Length
    Ext.Ø
    Availability
    Price
  • 5
    0,41 mm
    22
    51 mm
    0,72 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 242,00 €
    242,00 €
    1 * 6 ST
       242,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Removable needle adapter, to convert a large hub removable needle termination into a Female Luer Lock termination
    Removable needle adapter, to convert a large hub removable needle termination into a Female Luer Lock termination
    HAMI35081
    Log in for availability
    1 * 1 ST - 59,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Removable needle adapter, to convert a large hub removable needle termination into a Male Luer Tip (LT) termination
    Removable needle adapter, to convert a large hub removable needle termination into a Male Luer Tip (LT) termination
    HAMI35080
    Log in for availability
    1 * 1 ST - 48,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Removable needle adapter, to convert a large hub removable needle termination into a Male Luer Lock (TLL) termination
    Removable needle adapter, to convert a large hub removable needle termination into a Male Luer Lock (TLL) termination
    HAMI35083
    Log in for availability
    1 * 1 ST - 53,00 €
Product Information
  • Catalog #
    Gauge
    Length
    Ext.Ø
    Availability
    Price
  • HAMI7784-02
    22
    51 mm
    0,72 mm
    Log in for availability
    1 * 6 ST - 242,00 €
Product Information
  • Catalog #
    Gauge
    Length
    Ext.Ø
    Availability
    Price
  • HAMI7779-01
    22
    51 mm
    0,72 mm
    Log in for availability
    1 * 6 ST - 83,30 €
Product Information
  • Catalog #
    Gauge
    Length
    Ext.Ø
    Availability
    Price
  • HAMI7784-01
    22s
    51 mm
    0,72 mm
    Log in for availability
    1 * 6 ST - 246,00 €
Product Information
  • Catalog #
    Gauge
    Length
    Ext.Ø
    Availability
    Price
  • HAMI7779-03
    22s
    51 mm
    0,72 mm
    Log in for availability
    1 * 6 ST - 83,30 €
Product Information
  • Catalog #
    Gauge
    Length
    Ext.Ø
    Availability
    Price
  • HAMI7780-03
    22s
    51 mm
    0,72 mm
    Log in for availability
    1 * 6 ST - 66,00 €
Product Information
  • Catalog #
    Gauge
    Length
    Ext.Ø
    Availability
    Price
  • HAMI7780-04
    22
    51 mm
    0,72 mm
    Log in for availability
    1 * 6 ST - 63,90 €