Flasks, flat bottom, wide neck, with beaded rim
Supplier: Lenz Laborglas
Jump to:
About this item
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area. Flasks with neck external diameter larger than 65 mm have reinforced rims.
Certifications: Acc. to DIN ISO 24450
6 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area. Flasks with neck external diameter larger than 65 mm have reinforced rims.
Certifications: Acc. to DIN ISO 24450
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Product Family Options
Product Information
- CapacityHeightNeck Ø ext.Ext.ØAvailabilityPrice