Orders LinkContactUsLinkComponent
Dissolved oxygen probes, InLab® OptiOx™
Dissolved oxygen probes, InLab® OptiOx™
Supplier:   Mettler - Toledo
Dissolved oxygen probes, InLab® OptiOx™
Supplier:  Mettler - Toledo
Certificates
Dissolved oxygen probes, InLab® OptiOx™
Dissolved oxygen probes, InLab® OptiOx™
3 Options Available Below
Product Details & Documents
The InLab® OptiOx optical dissolved oxygen sensor is based on the reliable RDO® (Rugged Dissolved Oxygen) technology. RDO® is fast, precise and produces extremely reproducible results.
  • Stable results and rapid response time
  • Immediately ready for use
  • Extremely easy handling, zero maintenance: save time
  • Suitable for an extremely wide range of applications
The InLab® OptiOx sensors can be used in combination with a SevenExcellence benchtop meter or the SevenGo 9 or SevenGo Duo 98 portable meters.
Documents
Product Family Options
Product Information
  • Type
    Concentration
    Temp.
    Connection
    Ø×L
    Availability
    Price
  • 664-0075
    Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    InLab® OptiOx™
    0 - 50 mg/l (0 - 500%)
    0...+50 °C
    Mini LTW, 1,8 m fixed cable
    16×65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 650,00 €
    1 650,00 €
    1 * 1 ST
       1 650,00 €
  • 664-0077
    Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    InLab® OptiOx™
    0 - 50 mg/l (0 - 500%)
    0...+50 °C
    Mini LTW, 10 m fixed cable
    16×65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 810,00 €
    1 810,00 €
    1 * 1 ST
       1 810,00 €
  • 664-0076
    Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    InLab® OptiOx™
    0 - 50 mg/l (0 - 500%)
    0...+50 °C
    Mini LTW, 5 m fixed cable
    16×65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 580,00 €
    1 580,00 €
    1 * 1 ST
       1 580,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Zero oxygen tablets, used for calibration
    Zero oxygen tablets, used for calibration
    METT51300140
    Log in for availability
    1 * 20 ST - 53,50 €