Order Entry
Belgium
ContactUsLinkComponent
Multi-parameter meter (pH/ion/oxygen), handheld, SevenGo Duo pro™ SG68
Multi-parameter meter (pH/ion/oxygen), handheld, SevenGo Duo pro™ SG68
Supplier:  Mettler - Toledo
Certificates
undefined
Multi-parameter meter (pH/ion/oxygen), handheld, SevenGo Duo pro™ SG68
Supplier:  Mettler - Toledo

Specifications

  • Catalog No:
  • 665-0194
  • 665-0193
  • 665-0268
  • 665-0267
  • Model:
  • SG68
  • SG68
  • SG68
  • SG68
  • pH range:
  • –2,000...+19,999
  • –2,000...+19,999
  • –2,000...+19,999
  • –2,000...+19,999
  • pH resolution:
  • 0,001
  • 0,001
  • 0,001
  • 0,001
  • pH accuracy:
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • pH calibration:
  • Max. 5 points, 7 pre-defined buffer groups
  • Max. 5 points, 7 pre-defined buffer groups
  • Max. 5 points, 7 pre-defined buffer groups
  • Max. 5 points, 7 pre-defined buffer groups
  • mV range:
  • ±1999
  • ±1999
  • ±1999
  • ±1999
  • mV resolution:
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • mV accuracy:
  • ±0,1
  • ±0,1
  • ±0,1
  • ±0,1
  • ISE range:
  • 1,00×10-9 - 9,99×10+9 / 0,000 - 9999 ppm / 0,000 - 999,9%
  • 1,00×10-9 - 9,99×10+9 / 0,000 - 9999 ppm / 0,000 - 999,9%
  • 1,00×10-9 - 9,99×10+9 / 0,000 - 9999 ppm / 0,000 - 999,9%
  • 1,00×10-9 - 9,99×10+9 / 0,000 - 9999 ppm / 0,000 - 999,9%
  • ISE accuracy:
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ISE units:
  • %, ppm, mg/l, mmol/l
  • %, ppm, mg/l, mmol/l
  • %, ppm, mg/l, mmol/l
  • %, ppm, mg/l, mmol/l
  • DO concentration:
  • 0,00 - 99,00 mg/L (0,0 - 600%)
  • 0,00 - 99,00 mg/L (0,0 - 600%)
  • 0,00 - 99,00 mg/L (0,0 - 600%)
  • 0,00 - 99,00 mg/L (0,0 - 600%)
  • DO concentration resolution:
  • 0,01 mg/L (0,1 / 1%)
  • 0,01 mg/L (0,1 / 1%)
  • 0,01 mg/L (0,1 / 1%)
  • 0,01 mg/L (0,1 / 1%)
  • DO accuracy:
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • Temperature range:
  • 0,0...+60,0 °C
  • 0,0...+60,0 °C
  • 0,0...+60,0 °C
  • 0,0...+60,0 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • Power supply:
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • Compliance:
  • IP 67/GLP
  • IP 67/GLP
  • IP 67/GLP
  • IP 67/GLP
  • Data logging capacity:
  • 500 data records
  • 500 data records
  • 500 data records
  • 500 data records
  • Outputs:
  • IR to printer or PC via RS232 or USB
  • IR to printer or PC via RS232 or USB
  • IR to printer or PC via RS232 or USB
  • IR to printer or PC via RS232 or USB
  • W×D×H:
  • 220×90×45 mm
  • 220×90×45 mm
  • 220×90×45 mm
  • 220×90×45 mm
  • Weight:
  • 0,368 kg (without battery)
  • 0,368 kg (without battery)
  • 0,368 kg (without battery)
  • 0,368 kg (without battery)

Specifications

Related Information

Oxygen probe

Product Information

Frequently Bought Together