Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Multi-channel pipettes, mechanical, variable volume, SoftGrip™
Multi-channel pipettes, mechanical, variable volume, SoftGrip™
Supplier:   HAMILTON BONADUZ
Multi-channel pipettes, mechanical, variable volume, SoftGrip™
Supplier:  HAMILTON BONADUZ
Certificates
Multi-channel pipettes, mechanical, variable volume, SoftGrip™
Multi-channel pipettes, mechanical, variable volume, SoftGrip™
4 Options Available Below
Product Details & Documents
Hamilton SoftGrip pipettes are available as 8-channel and 12-channel adjustable volume manual pipettes in two volume ranges covering 5 to 50 µl and 30 to 300 µl.
  • Long tip ejector lever and movable lower body can be rotated a full 360º relative to the handle to provide maximum comfort for left- or right-handed use
  • Colour-coded plunger buttons and labels for quick volume identification
  • Easily activated tip ejector lever releases all the tips in single stroke
  • Can be autoclaved without the need for disassembly or recalibration
Please note: Accuracy and precision is guaranteed when used with Hamilton Universal AdvanTip™ precision pipette tips.
Product Family Options
8-Channel
Product Information
  • Volume
    Accroissement
    Précision
    Imprécision
    Code couleur
    Pour pointe
    Availability
    Price
  • 5 - 50 µl
    0,02
    ±0,8 à ±2,0 %
    ±0,35 - ±2,0 %
    marron
    200 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 858,00 €
    858,00 €
    1 * 1 ST
       858,00 €
  • 30 - 300 µl
    0,2
    ±0,5 à ±1,2 %
    ±0,3 - ±0,7 %
    rouge
    300 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 804,00 €
    804,00 €
    1 * 1 ST
       804,00 €
12-Channel
Product Information
  • Volume
    Accroissement
    Précision
    Imprécision
    Code couleur
    Pour pointe
    Availability
    Price
  • 5 - 50 µl
    0,02
    ±1,0 à ±5,0 %
    ±0,5 - ±2,0 %
    marron
    200 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 962,00 €
    962,00 €
    1 * 1 ST
       962,00 €
  • 30 - 300 µl
    0,2
    ±1,0 à ±2,4 %
    ±0,5 - ±1,2 %
    rouge
    300 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 998,00 €
    998,00 €
    1 * 1 ST
       998,00 €
Accessories
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • HAMI1987-01
    Labels d'étalonnage pour pipettes
    Log in for availability
    1 * 10 ST - 34,70 €
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • HAMI34002
    Clé d'étalonnage de pipettes, pour un étalonnage facile sur le terrain
    Log in for availability
    1 * 1 ST - 9,80 €
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • HAMI54756-01
    Ejector sleeve for SoftGrip™ single channel pipette, 500 - 1000 µl
    Log in for availability
    1 * 1 ST - 23,80 €